Order Entry
Export
Orders LinkContactUsLinkComponent
Pierce™ Avidin Agarose

Pierce™ Avidin Agarose

Avidin agarose, 5 ml
Catalog # PIER20219
Pierce™ Avidin Agarose
Catalog # PIER20219
CAS Number:  
Pierce™ Avidin Agarose
Pierce™ Avidin Agarose

Avidin agarose, 5 ml

CAS Number:  
Price
322,00 €
Log in for availability
1 * 5 mL
Product Details & Documents

Pierce™ Avidin agarose is purified avidin immobilised on crosslinked 6% beaded agarose for the affinity purification of biotinylated macromolecules.

  • Strong, nearly irreversible biotin:avidin interaction
  • Approximate binding capacity >20 µg biotin/ml resin
Specifications

Specifications

  • Pk:
    5 ml
  • Description:
    Avidin agarose

Specifications