Order Entry
Export
Orders LinkContactUsLinkComponent
Pierce™ DNA Extraction Buffer, Phenol:Chloroform and Tris Buffer

Pierce™ DNA Extraction Buffer, Phenol:Chloroform and Tris Buffer

Phenol:chloroform and tris buffer, 400 ml
Catalog # PIER17909
Pierce™ DNA Extraction Buffer, Phenol:Chloroform and Tris Buffer
Catalog # PIER17909
CAS Number:  
Pierce™ DNA Extraction Buffer, Phenol:Chloroform and Tris Buffer
Pierce™ DNA Extraction Buffer, Phenol:Chloroform and Tris Buffer

Phenol:chloroform and tris buffer, 400 ml

CAS Number:  
Price
301,00 €
Log in for availability
1 * 400 mL
Product Details & Documents

Phenol-chloroform, is phenol supplied premixed with chloroform:isoamyl alcohol (25:24:1) and accompanying bottle of buffer to equilibrate at either pH 6,7 or pH 8,0.

  • Offers excellent, nuclease-free performance
  • Ideal for lower pH DNA extractions and for most DNA applications
Specifications

Specifications

  • Pk:
    400 ml
  • Description:
    Phenol:chloroform and tris buffer

Specifications