Agarose I, Pierce™
Agarose I
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Agarose I (molecular biology grade) is a highly purified and carefully blended formulation that provides uniform lot-to-lot consistency for preparation of electrophoresis gels.
- Standard melting temperature agarose for electrophoresis
- Separation range: 100 bp to >30 kb
Documents
Certificates
Search by Lot NumberSpecifications
- Description:Agarose I