Top tilt measures
25 ml
Catalog # 612-9856
Supplier: Lenz Laborglas
Jump to:
Product Details & Documents
DURAN®, borosilicate glass 3.3, clear.
- Very good chemical resistance
- High temperature resistance
- Minimal thermal expansion giving relatively high resistance to temperature changes
These tilt measures with ground joint cone NS 29/32 can fit flasks - see 215-1117 and 215-1118.
Documents
BSE TSE Glass Columns
BSE/TSE Certificates
Specifications
- Capacity:25 ml