Orders LinkContactUsLinkComponent
Top tilt measures
Top tilt measures
10 ml
Catalog # 612-9853
Supplier:   Lenz Laborglas
Top tilt measures
Catalog # 612-9853
Supplier:  Lenz Laborglas
Top tilt measures
Top tilt measures
10 ml
Price
47,60 €
Log in for availability
1 * 1 items
Product Details & Documents
DURAN®, borosilicate glass 3.3, clear.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
These tilt measures with ground joint cone NS 29/32 can fit flasks - see 215-1117 and 215-1118.
Documents
Specifications
  • Capacity:
    10 ml