Flasks, pear shaped with two necks
50 ml, 14/23 NS, 14/23 NS, 50 mm
271-1233
DURAN®, borosilicate glass.
- Excellent chemical resistance
- High temperature resistance
- Minimal thermal expansion, giving relatively high resistance to temperature changes
The conical geometry makes them ideal for small-scale reactions.
- Ext.Ø:50 mm
- Capacity:50 ml
- Side necks:14/23 NS
- Centre neck:14/23 NS



