Orders LinkContactUsLinkComponent
Adapters, expansion, DURAN®
Adapters, expansion, DURAN®
Expansion, 24/29 NS, 29/32 NS
Catalog # SCOT286548201
Supplier:   DWK Life Sciences
Adapters, expansion, DURAN®
Catalog # SCOT286548201
Supplier:  DWK Life Sciences
Adapters, expansion, DURAN®
Adapters, expansion, DURAN®
Expansion, 24/29 NS, 29/32 NS
Price
223,00 €
Log in for availability
1 * 10 items
Product Details & Documents
DURAN®, borosilicate glass 3,3 clear.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Expansion adapter from small cone to large socket.
Specifications
  • Cone size:
    24/29 NS
  • Socket size:
    29/32 NS
  • Type:
    Expansion
  • Seq:
    07