Stopcocks, single way, DURAN®
4 mm, 14,5 NS
Catalog # SCOT286439807
Supplier: DWK Life Sciences
Jump to:
Product Details & Documents
DURAN®, borosilicate glass 3,3, clear, polished, without key.
- Very good chemical resistance
- Very good temperature resistance
- For glass manipulators
Ungrounded, without key.
Specifications
- Hole Ø:4 mm
- Spigot:14,5 NS