Orders LinkContactUsLinkComponent
Stopcocks, single way, DURAN®
Stopcocks, single way, DURAN®
2,5 mm, 14,5 NS
Catalog # SCOT286439601
Supplier:   DWK Life Sciences
Stopcocks, single way, DURAN®
Catalog # SCOT286439601
Supplier:  DWK Life Sciences
Stopcocks, single way, DURAN®
Stopcocks, single way, DURAN®
2,5 mm, 14,5 NS
Price
848,00 €
Log in for availability
1 * 50 items
Product Details & Documents
DURAN®, borosilicate glass 3,3, clear, polished, without key.
  • Very good chemical resistance
  • Very good temperature resistance
  • For glass manipulators
Ungrounded, without key.
Specifications
  • Hole Ø:
    2,5 mm
  • Spigot:
    14,5 NS