Orders LinkContactUsLinkComponent
Stopcocks for burettes, angled, DURAN®
Stopcocks for burettes, angled, DURAN®
with SBW glass key, 2,5 mm, 12,5 NS
Catalog # SCOT286409204
Supplier:   DWK Life Sciences
Stopcocks for burettes, angled, DURAN®
Catalog # SCOT286409204
Supplier:  DWK Life Sciences
Stopcocks for burettes, angled, DURAN®
Stopcocks for burettes, angled, DURAN®
with SBW glass key, 2,5 mm, 12,5 NS
Price
1 190,00 €
Log in for availability
1 * 50 items
Product Details & Documents
DURAN®, borosilicate glass 3.3, with glass key.
  • Very good chemical resistance
  • Very good temperature resistance
  • Screwthread retaining device
Specifications
  • Hole Ø:
    2,5 mm
  • Description:
    with SBW glass key
  • Spigot:
    12,5 NS