Stopcocks, single-way, DURAN®
2,5 mm, 12,5 NS
Catalog # SCOT286403604
Supplier: DWK Life Sciences
Jump to:
Product Details & Documents
DURAN®, borosilicate glass 3.3, clear, with glass or PTFE keys.
- Replaceable NS keys
- Screw thread retaining device
Straight bore.
Specifications
- Hole Ø:2,5 mm
- Seq:08
- Spigot:12,5 NS