Screwthread tubes, attachable, DURAN®
GL 32, 28 mm, 2,0 mm
SCOT286258808
DURAN®, borosilicate glass 3.3, clear.
- Very good chemical resistance
- High temperature resistance
- Minimal thermal expansion giving relatively high resistance to temperature changes
Screw thread GL. It is used by glassblowers in the construction of laboratory glassware.
: These tubes are open at the bottom.
- Thread:GL 32
- Tube ext.Ø:28 mm
- Wall thickness:2,0 mm
- Seq:04



