Order Entry
Export
Orders LinkContactUsLinkComponent
Electrolyte solutions, organic

Electrolyte solutions, organic

Electrolyte solution, L 5014, LiCl saturated in glacial acetic acid, 250 ml in DURAN® glass bottle, 250 mL
Catalog # SCOI285138324
Supplier:   SI Analytics
Electrolyte solutions, organic
Catalog # SCOI285138324
Supplier:  SI Analytics
CAS Number:  
Electrolyte solutions, organic
Electrolyte solutions, organic

Electrolyte solution, L 5014, LiCl saturated in glacial acetic acid, 250 ml in DURAN® glass bottle, 250 mL

CAS Number:  
Price
119,00 €
Log in for availability
1 * 250 mL
Product Details & Documents

Electrolyte solutions for measurements in organic solutions for reference electrodes and as electrolyte bridges.

Specifications

Specifications

  • Pk:
    250 mL
  • Description:
    Electrolyte solution, L 5014, LiCl saturated in glacial acetic acid, 250 ml in DURAN® glass bottle

Specifications