Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
ASTM solid-stem thermometer GOLDBRAND yellow, ASTM No. 22 C + 95 +103C/0,1C suit.cert. 1 * 1 items
Catalog # BRND880021
Supplier:
Brand
Expand image
ASTM solid-stem thermometer GOLDBRAND yellow, ASTM No. 22 C + 95 +103C/0,1C suit.cert. 1 * 1 items
Catalog # BRND880021
Supplier:
Brand