Digital Solutions
Applications
Support
About
Export
ContactUsLinkComponent
Thermometer encl.-scale SILBERBRAND total immersion, -10 +420C subdiv. 1C opal glass scale 1 * 1 items
BRND800409
:
Brand
Thermometer encl.-scale SILBERBRAND total immersion, -10 +420C subdiv. 1C opal glass scale 1 * 1 items
BRND800409
:
Brand