Order Entry
Export
Orders LinkContactUsLinkComponent
Tubes, rack packed, non-sterile

Tubes, rack packed, non-sterile

Individual tubes, PP
Catalog # 211-3254
Supplier:   Brand
Tubes, rack packed, non-sterile
Catalog # 211-3254
Supplier:  Brand
Tubes, rack packed, non-sterile
Tubes, rack packed, non-sterile

Individual tubes, PP

Price
75,40 €
Log in for availability
1 * 960 items
Product Details & Documents

PP tubes and PE caps, non-sterile.

  • Tubes and racks are autoclavable at 121 °C (2 bar) according to DIN EN 285 (not caps)
  • Designed for storing and freezing down to −80 °C and for transporting reagents and samples
  • Compatible with standard microplates
  • Suitable for working with multi-channel pipettors

Suitable for biological tests, such as PCR, cell uptake studies, RIA and EIA.

Tube o. Ø×h: 8,8×45 mm

Specifications
  • Description:
    Individual tubes
  • Material:
    PP