Order Entry
Export
Orders LinkContactUsLinkComponent
 

Tubes, rack packed, non-sterile

Rack with 96 individual 1,2 ml tubes
  211-3252
 :   Brand
Tubes, rack packed, non-sterile
  211-3252
 :  Brand
Tubes, rack packed, non-sterile
Tubes, rack packed, non-sterile

Rack with 96 individual 1,2 ml tubes

 
122,00 €
   
1 * 10 items
 

PP tubes and PE caps, non-sterile.

  • Tubes and racks are autoclavable at 121 °C (2 bar) according to DIN EN 285 (not caps)
  • Designed for storing and freezing down to −80 °C and for transporting reagents and samples
  • Compatible with standard microplates
  • Suitable for working with multi-channel pipettors

Suitable for biological tests, such as PCR, cell uptake studies, RIA and EIA.

Tube o. Ø×h: 8,8×45 mm

 
  • Description:
    Rack with 96 individual 1,2 ml tubes