Orders LinkContactUsLinkComponent
Staining troughs with lid
Staining troughs with lid
Wire handle, to move tray
Catalog # BRND473100
Supplier:   Brand
Staining troughs with lid
Catalog # BRND473100
Supplier:  Brand
Staining troughs with lid
Staining troughs with lid
Wire handle, to move tray
Price
42,50 €
Log in for availability
1 * 10 items
Product Details & Documents
Soda-lime glass.
For 10 standard slides, size (76×26 mm)
Ordering information: Please order staining trough, tray, and wire handle seperately.
Specifications
  • Description:
    Wire handle, to move tray