Staining troughs with lid
Staining troughs, with lid, 85×105×70 mm
Catalog # BRND472200
Supplier: Brand
Jump to:
Product Details & Documents
Soda-lime glass.
For 10 standard slides, size (76×26 mm)
: Please order staining trough, tray, and wire handle seperately.
Specifications
- Description:Staining troughs, with lid
- W×D×H:85×105×70 mm



