Orders LinkContactUsLinkComponent
Density bottles, uncalibrated
Density bottles, uncalibrated
100 ml
Catalog # 614-0344
Supplier:   Brand
Density bottles, uncalibrated
Catalog # 614-0344
Supplier:  Brand
Density bottles, uncalibrated
Density bottles, uncalibrated
100 ml
Price
117,00 €
Log in for availability
1 * 2 items
Product Details & Documents
Borosilicate 3.3 glass, Gay-Lussac type
  • Stopper NS 10/19 with capillary
  • Top of stopper ground and polished
  • Nominal capacity printed on the bottom
Certifications: DIN ISO 3507
Specifications
  • Volume (nom.):
    100 ml