Density bottles, uncalibrated
50 ml
614-0343
: Brand
Jump to:
Borosilicate 3.3 glass, Gay-Lussac type
- Stopper NS 10/19 with capillary
- Top of stopper ground and polished
- Nominal capacity printed on the bottom
Certifications: DIN ISO 3507
Specifications
- Volume (nom.):50 ml