Order Entry
Export
Orders LinkContactUsLinkComponent
Automatic burettes, Pellet pattern, without intermediate stopcock, Schellbach stripe, SILBERBRAND

Automatic burettes, Pellet pattern, without intermediate stopcock, Schellbach stripe, SILBERBRAND

25 ml, 0,05 ml, 0,05 ± ml
Catalog # BRND21912
Supplier:   Brand
Automatic burettes, Pellet pattern, without intermediate stopcock, Schellbach stripe, SILBERBRAND
Catalog # BRND21912
Supplier:  Brand
Automatic burettes, Pellet pattern, without intermediate stopcock, Schellbach stripe, SILBERBRAND
Automatic burettes, Pellet pattern, without intermediate stopcock, Schellbach stripe, SILBERBRAND

25 ml, 0,05 ml, 0,05 ± ml

Price
199,00 €
Log in for availability
1 * 1 items
Product Details & Documents

Borosilicate glass 3.3, class B.

  • Calibrated to deliver 'TD, Ex'
  • Schellbach stripe, ring marks at major graduations
  • Automatic zeroing, needle-valve stopcock with PTFE spindle, without intermediate stopcock

Certifications: DIN EN ISO 385

Specifications
  • Capacity:
    25 ml
  • Division:
    0,05 ml
  • Tolerance:
    0,05 ± ml
Accessories

Product Information

  • Catalog #
    Capacity
    Availability
    Price
  • Reservoir bottle, clear glass
    Reservoir bottle, clear glass
    612-3219
    2000 ml
    Log in for availability
    1 * 1 items - 74,30 €

Product Information

  • Catalog #
    Capacity
    Availability
    Price
  • Ball set with net, black, size 5, natural rubber/PVC
    Ball set with net, black, size 5, natural rubber/PVC
    612-9953
    102
    Log in for availability
    1 * 1 items - 107,00 €

Product Information

  • Catalog #
    Availability
    Price
  • Single bulb rubber bellows, orange coloured, with valve and tubing
    Single bulb rubber bellows, orange coloured, with valve and tubing
    BRND23400
    Log in for availability
    1 * 1 items - 16,10 €

Product Information

  • Catalog #
    Capacity
    Availability
    Price
  • Reservoir bottle, clear soda-lime glass, NS29/32
    Reservoir bottle, clear soda-lime glass, NS29/32
    BRND126965
    2000 ml
    Log in for availability
    1 * 1 items - 30,80 €

Product Information

  • Catalog #
    Hole Ø
    Availability
    Price
  • Spare spindle, for 2-50 ml burette capacity
    Spare spindle, for 2-50 ml burette capacity
    BRND82170
    0 - 2,5 mm
    Log in for availability
    1 * 1 items - 18,10 €