Order Entry
Export
Orders LinkContactUsLinkComponent
Diethyl ether stabilised, BAKER ANALYZED® ACS, J.T.Baker®

Diethyl ether stabilised, BAKER ANALYZED® ACS, J.T.Baker®

Glass bottle, 1 L
Catalog # 8254.1000
Flammable
Irritant
Supplier:   Avantor
Diethyl ether stabilised, BAKER ANALYZED® ACS, J.T.Baker®
Catalog # 8254.1000
Supplier:  Avantor
CAS Number:  60-29-7
Diethyl ether stabilised, BAKER ANALYZED® ACS, J.T.Baker®
Diethyl ether stabilised, BAKER ANALYZED® ACS, J.T.Baker®

Glass bottle, 1 L

CAS Number:  60-29-7
Price
75,30 €
Log in for availability
1 * 1 L
Product Details & Documents
Synonyms:  Ethyl Ether, Ether, 2,2'-Oxydiethane, 2,2'-Oxybisethane, Ethoxyethane
  • CAS Number:60-29-7
  • Formula:(CH₃CH₂)₂O
  • MDL Number:MFCD00011646
  • Molecular Weight:74.12 g/mol
  • Merck Index:12,03852
  • Boiling Pt:35 °C (1013 hPa)
  • Melting Pt:–123 °C
  • Density:0.7135 g/cm³ (20 °C)
  • Flash Pt:–40 °C
  • Storage Temperature:Ambient
  • EINECS:200-467-2
  • UN:1155
  • ADR:3,I
  • REACH:01-2119535785-29
Specifications

Specification Test Results

  • Assay (by GC):
    min. 99,5 %
  • Alcohol (C2H5OH):
    max. 0,01 %
  • Carbonyl Compounds (as HCHO):
    max. 0,001 %
  • Color (APHA):
    max. 10
  • Density (g/ml) at 20°C:
    0.712-0.714
  • Foreign Odor:
    passes test
  • Peroxide (as H202):
    max. 3 ppm
  • Residue after Evaporation:
    max. 0,001 %
  • Substances Darkened by H2SO4:
    passes test
  • Titrable Acid (meq/g):
    max. 0,0002
  • Water (H2O):
    max. 0,01 %
  • Trace Impurities (in ppm):
    -
  • Aluminium (Al):
    max. 0,5
  • Barium (Ba):
    max. 0,1
  • Boron (B):
    max. 0,02
  • Cadmium (Cd):
    max. 0,05
  • Calcium (Ca):
    max. 0,5
  • Chromium (Cr):
    max. 0,02
  • Cobalt (Co):
    max. 0,02
  • Copper (Cu):
    max. 0,02
  • Iron (Fe):
    max. 0,1
  • Lead (Pb):
    max. 0,1
  • Magnesium (Mg):
    max. 0,1
  • Manganese (Mn):
    max. 0,02
  • Nickel (Ni):
    max. 0,02
  • Tin (Sn):
    max. 0,1
  • Zinc (Zn):
    max. 0,1

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L

Specifications