Order Entry
Export
Orders LinkContactUsLinkComponent
Cyclohexane, BAKER ANALYZED® ACS, J.T.Baker®

Cyclohexane, BAKER ANALYZED® ACS, J.T.Baker®

25 L
Catalog # 8026.9025
Flammable
Health
Irritant
Environment
Supplier:   Avantor
Cyclohexane, BAKER ANALYZED® ACS, J.T.Baker®
Catalog # 8026.9025
Supplier:  Avantor
CAS Number:  110-82-7
Cyclohexane, BAKER ANALYZED® ACS, J.T.Baker®
Cyclohexane, BAKER ANALYZED® ACS, J.T.Baker®

25 L

CAS Number:  110-82-7
Price
Inquire For Price
Log in for availability
1 * 25 L
Product Details & Documents
Synonyms:  Hexahydrobenzene, Hexamethylene
  • CAS Number:110-82-7
  • Formula:C₆H₁₂
  • MDL Number:MFCD00003814
  • Molecular Weight:84.16 g/mol
  • Merck Index:13,02752
  • Boiling Pt:81 °C (1013 hPa)
  • Melting Pt:7 °C
  • Density:0.7785 g/cm³ (20 °C)
  • Flash Pt:–18 °C
  • Storage Temperature:Ambient
  • EINECS:203-806-2
  • UN:1145
  • ADR:3,II
Specifications

Specification Test Results

  • Assay (by GC):
    min. 99,0 %
  • Benzene (C6H6):
    max. 0,005 %
  • Color (APHA):
    max. 10
  • Residue after Evaporation:
    max. 0,002 %
  • Substances Darkened by H2SO4:
    passes test
  • Water (H2O):
    max. 0,02 %
  • Trace Impurities (in ppm):
    -
  • Aluminium (Al):
    max. 0,5
  • Barium (Ba):
    max. 0,1
  • Boron (B):
    max. 0,02
  • Cadmium (Cd):
    max. 0,05
  • Calcium (Ca):
    max. 0,5
  • Chromium (Cr):
    max. 0,02
  • Cobalt (Co):
    max. 0,02
  • Copper (Cu):
    max. 0,02
  • Iron (Fe):
    max. 0,1
  • Lead (Pb):
    max. 0,1
  • Magnesium (Mg):
    max. 0,1
  • Manganese (Mn):
    max. 0,02
  • Nickel (Ni):
    max. 0,02
  • Tin (Sn):
    max. 0,1
  • Zinc (Zn):
    max. 0,1

Specifications

  • Pk:
    25 L

Specifications