Cyclohexane, BAKER ANALYZED® ACS, J.T.Baker®
Glass bottle, 2,5 L
: Avantor
: Hexahydrobenzene, Hexamethylene
- :110-82-7
- :C₆H₁₂
- :MFCD00003814
- :84.16 g/mol
- :13,02752
- :81 °C (1013 hPa)
- :7 °C
- :0.7785 g/cm³ (20 °C)
- :–18 °C
- :Ambient
- :203-806-2
- :1145
- :3,II
- Assay (by GC):min. 99,0 %
- Benzene (C6H6):max. 0,005 %
- Color (APHA):max. 10
- Residue after Evaporation:max. 0,002 %
- Substances Darkened by H2SO4:passes test
- Water (H2O):max. 0,02 %
- Trace Impurities (in ppm):-
- Aluminium (Al):max. 0,5
- Barium (Ba):max. 0,1
- Boron (B):max. 0,02
- Cadmium (Cd):max. 0,05
- Calcium (Ca):max. 0,5
- Chromium (Cr):max. 0,02
- Cobalt (Co):max. 0,02
- Copper (Cu):max. 0,02
- Iron (Fe):max. 0,1
- Lead (Pb):max. 0,1
- Magnesium (Mg):max. 0,1
- Manganese (Mn):max. 0,02
- Nickel (Ni):max. 0,02
- Tin (Sn):max. 0,1
- Zinc (Zn):max. 0,1
- Pack type:Glass bottle
- Pk:2,5 L



