Order Entry
Export
Orders LinkContactUsLinkComponent
 

Acetone, BAKER, J.T.Baker®

5 L
  8003.5000
 
 
   Safer Human and Environmental Health
 :   Avantor
Acetone, BAKER, J.T.Baker®
  8003.5000
 :  Avantor
   Safer Human and Environmental Health
 :  67-64-1
Acetone, BAKER, J.T.Baker®
Acetone, BAKER, J.T.Baker®

5 L

 :  67-64-1
 
153,32 €
   
1 * 5 L
 
 :  2-Propanone, DMK, Propanone, Dimethyl ketone
  •  :67-64-1
  •  :CH₃COCH₃
  •  :MFCD00008765
  •  :58.08 g/mol
  •  :13,00067
  •  :56.2 °C (1013 hPa)
  •  :–95.4 °C
  •  :0.792 g/cm³ (20 °C)
  •  :<–20 °C (closed cup)
  •  :Ambient
  •  :200-662-2
  •  :1090
  •  :3,II
  •  :01-2119471330-49
 

 

  • Acidity or Alkalinity:
    passes test
  • Appearance of solution:
    passes test
  • Identification:
    passes test
  • Matter Insoluble in Water:
    passes test
  • Reducing Substances:
    passes test
  • Relative Density:
    0.790-0.793
  • Residue on Evaporation:
    max. 50 ppm
  • Water (H2O):
    max. 3 g/l
  • Additional Specification(s)::
    -
  • Assay (by GC):
    min. 99,0 %
  • Related Substances::
    -
  • Benzene:
    max. 2 ppm (v/v)
  • Methanol (CH3OH):
    max. 0,05 % (v/v)
  • Other Impurities:
    passes test
  • 2-Propanol (CH3CHOHCH3):
    max. 0,05 % (v/v)

 

  • Pk:
    5 L
  • Cat. No.:
    8003.5000
  • Environmental certification:
    Chem21 Solvent Selection Guide recommended after discussion
  • Environmentally Preferable:
    Safer Human and Environmental Health