Acetone, BAKER, J.T.Baker®
Glass bottle, 1 L
Supplier: Avantor
Acetone, BAKER, J.T.Baker®
Catalog # 8003.1000
Supplier: Avantor
CAS Number: 67-64-1
Jump to:
Product Details & Documents
Synonyms: 2-Propanone, DMK, Propanone, Dimethyl ketone
- CAS Number:67-64-1
- Formula:CH₃COCH₃
- MDL Number:MFCD00008765
- Molecular Weight:58.08 g/mol
- Merck Index:13,00067
- Boiling Pt:56.2 °C (1013 hPa)
- Melting Pt:–95.4 °C
- Density:0.792 g/cm³ (20 °C)
- Flash Pt:<–20 °C (closed cup)
- Storage Temperature:Ambient
- EINECS:200-662-2
- UN:1090
- ADR:3,II
- REACH:01-2119471330-49
Documents
Certificates
Search by Lot NumberSpecifications
Specification Test Results
- Acidity or Alkalinity:passes test
- Appearance of solution:passes test
- Identification:passes test
- Matter Insoluble in Water:passes test
- Reducing Substances:passes test
- Relative Density:0.790-0.793
- Residue on Evaporation:max. 50 ppm
- Water (H2O):max. 3 g/l
- Additional Specification(s)::-
- Assay (by GC):min. 99,0 %
- Related Substances::-
- Benzene:max. 2 ppm (v/v)
- Methanol (CH3OH):max. 0,05 % (v/v)
- Other Impurities:passes test
- 2-Propanol (CH3CHOHCH3):max. 0,05 % (v/v)
Specifications
- Pack type:Glass bottle
- Pk:1 L
- Cat. No.:8003.1000
- Environmental certification:Chem21 Solvent Selection Guide recommended after discussion
- Environmentally Preferable:Safer Human and Environmental Health



