L(+)-Ascorbic acid, BAKER, Laboratory Reagent, J.T.Baker®
Plastic bottle for solids, 500 g
Supplier: Avantor
L(+)-Ascorbic acid, BAKER, Laboratory Reagent, J.T.Baker®
Catalog # 1914.0500
Supplier: Avantor
CAS Number: 50-81-7
Jump to:
Product Details & Documents
Synonyms: Ascorbic Acid, L-Ascorbic acid, (R)-5-((S)-1,2-Dihydroxyethyl)-3,4-dihydroxyfuran-2(5H)-one, Acidum ascorbicum
- CAS Number:50-81-7
- Formula:C₆H₈O₆
- MDL Number:MFCD00064328
- Molecular Weight:176.13 g/mol
- Merck Index:13,00837
- Boiling Pt:553 °C (1013 hPa)
- Melting Pt:190…192 °C
- Density:1.694 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:200-066-2
- REACH:exempt
Documents
Specifications
Specification Test Results
- Assay:99.0-100.5 %
- Appearance of solution:passes test
- Copper (Cu):max. 5 ppm
- Heavy Metals (as Pb):max. 10 ppm
- Identification:passes test
- Iron (Fe):max. 2 ppm
- Oxalic acid:passes test
- Specific Optical Rotation:+20.5 - +21.5 °
- Sulfated Ash :max. 0,10 %
Specifications
- Pack type:Plastic bottle for solids
- Pk:500 g



