Order Entry
Export
Orders LinkContactUsLinkComponent
L(+)-Ascorbic acid, BAKER ANALYZED® for biotechnology, J.T.Baker®

L(+)-Ascorbic acid, BAKER ANALYZED® for biotechnology, J.T.Baker®

Glass bottle for solids, 500 g
Catalog # 1018.0500
Supplier:   Avantor
L(+)-Ascorbic acid, BAKER ANALYZED® for biotechnology, J.T.Baker®
Catalog # 1018.0500
Supplier:  Avantor
CAS Number:  50-81-7
L(+)-Ascorbic acid, BAKER ANALYZED® for biotechnology, J.T.Baker®
L(+)-Ascorbic acid, BAKER ANALYZED® for biotechnology, J.T.Baker®

Glass bottle for solids, 500 g

CAS Number:  50-81-7
Price
266,80 €
Log in for availability
1 * 500 g
Product Details & Documents
Synonyms:  Ascorbic Acid, L-Ascorbic acid, (R)-5-((S)-1,2-Dihydroxyethyl)-3,4-dihydroxyfuran-2(5H)-one, Acidum ascorbicum
  • CAS Number:50-81-7
  • Formula:C₆H₈O₆
  • MDL Number:MFCD00064328
  • Molecular Weight:176.13 g/mol
  • Merck Index:13,00837
  • Boiling Pt:553 °C (1013 hPa)
  • Melting Pt:190…192 °C
  • Density:1.694 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:200-066-2
  • REACH:exempt

Documents

Specifications

Specification Test Results

  • Assay (dried):
    min. 99,5 %
  • Heavy Metals (as Pb):
    max. 0,001 %
  • Iron (Fe):
    max. 5 ppm
  • Loss on Drying at 105°C:
    max. 0,1 %
  • Residue on Ignition:
    max. 0,05 %
  • Specific Rotation [alpha] 20/D (dried basis), c=10 in water:
    +20.5 - +21.5 °
  • Water Insoluble Material:
    passes test

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    500 g

Specifications