Orders LinkContactUsLinkComponent
Burette clamps
0 - 20 mm, One burette
  241-7196
 :   BOCHEM
Burette clamps
  241-7196
 :  BOCHEM
Burette clamps
Burette clamps
0 - 20 mm, One burette
54,00 €
1 * 1 items
Stainless steel. For simple and secure fixing of burettes.
  • With bosshead for fixing to retort stand
  • Clamping range 0 to 20 mm
  • Jaws are covered with a silicone coating to protect the burettes
  • For:
    One burette
  • Spanning width:
    0 - 20 mm