Order Entry
Export
Orders LinkContactUsLinkComponent
Tangential flow cassettes, Centramate™

Tangential flow cassettes, Centramate™

Supor®, 0,2 µm, 1000 cm²
Catalog # 516-9698
Tangential flow cassettes, Centramate™
Catalog # 516-9698
Supplier:  
Tangential flow cassettes, Centramate™
Tangential flow cassettes, Centramate™

Supor®, 0,2 µm, 1000 cm²

Discontinued
Product Details & Documents
Specifications
  • Filtration area:
    1000 cm²
  • Membrane:
    Supor®
  • Pore size:
    0,2 µm
  • Seq:
    04