Order Entry
Export
Orders LinkContactUsLinkComponent
Tangential flow cassettes, Centramate™

Tangential flow cassettes, Centramate™

Supor®, 0,1 µm, 1000 cm²
Catalog # 516-9693
Tangential flow cassettes, Centramate™
Catalog # 516-9693
Tangential flow cassettes, Centramate™
Tangential flow cassettes, Centramate™

Supor®, 0,1 µm, 1000 cm²

Price
1 730,00 €
Log in for availability
1 * 1 items
Product Details & Documents
Specifications
  • Filtration area:
    1000 cm²
  • Membrane:
    Supor®
  • Pore size:
    0,1 µm
  • Seq:
    03