Tissue processing cassette, Histosette II
Green
Catalog # 631-9674
Supplier: DELTALAB
Jump to:
Product Details & Documents
Made of acetal polymer.
- With 45° angle writing area
- Suitable for automatic labellers
- No need to break the lid and then put it in place, as it is supplied pre-mounted
- Can be easily opened and closed with just one hand
Documents
Certificates
Search by Lot NumberSpecifications
- Colour:Green
- Seq:05



