Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
Orders Link
ContactUsLinkComponent
Home
Microscope Objectives
Lenses for Stemi 2000
Lenses for Stemi 2000
Print
Lenses for Stemi 2000
Front lens, 60 mm
Catalog # 630-2398
Supplier:
ZEISS
Lenses for Stemi 2000
Catalog # 630-2398
Supplier:
ZEISS
Expand image
Return to Top
Lenses for Stemi 2000
Front lens, 60 mm
Specifications
Add to cart
Jump to:
Specifications
Price
Inquire For Price
Log in
for availability
Unit of Measure
1 * 1 items
-
+
Inquire For Price
Other Product Family Options Available
Return Policy
Product Details & Documents
Specifications
Magnification:
1,25×
Description:
Front lens
Seq:
003
Working distance:
60 mm