Orders LinkContactUsLinkComponent
Pipette tips, ART™
Pipette tips, ART™
ART™ 200, 200 µl, -, 50,7 mm, Bulk, 1000/bag
Catalog # 612-9748
   Low Manufacturing Impact
Pipette tips, ART™
Catalog # 612-9748
   Low Manufacturing Impact
Pipette tips, ART™
ART™ 200, 200 µl, -, 50,7 mm, Bulk, 1000/bag
Price
38,00 €
Log in for availability
1 * 1.000 items
Product Details & Documents
These non filtered tips are designed to fit virtually any micropipette. They are suitable for use when working with the most difficult samples, such as radioactive compounds, viscous materials, fragile cell lines, viruses, difficult proteins and PCR reagents.
  • Non sterile tips are certified to be free of RNase, DNase, DNA, ATP, endotoxins, pyrogens and PCR inhibitors
  • Sterile tips are certified to be free of RNase, DNase, DNA, ATP, bioburden, endotoxins, pyrogens and PCR inhibitors
  • Rack cover can easily be realigned from a traditional lift-off to a hinged rack for one-handed access to tips
  • Front latch closes and locks ensure safe and secure storage
  • Space saving reload system is environmentally friendly, traceable and easy to use
Packaging: Tips are available in bulk packaging, in racks or in trays for rack reloading. Reload tips require an empty rack.
Specifications
  • Packed:
    Bulk, 1000/bag
  • Sterile:
    -
  • Volume:
    200 µl
  • Length:
    50,7 mm
  • Seq:
    14
  • Tip style:
    ART™ 200
  • Cat. No.:
    612-9748
  • Environmental certification:
    IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
  • Environmentally Preferable:
    Low Manufacturing Impact
Accessories
Product Information
  • Catalog #
    Availability
    Price
  • Empty hinged racks, medium, for 20 μl, 200 and 300 μl tips
    Empty hinged racks, medium, for 20 μl, 200 and 300 μl tips
    732-1170
    Log in for availability
    1 * 10 items - 43,90 €
Product Information
  • Catalog #
    Availability
    Price
  • Empty hinged racks, large, for 1000 μl tips
    Empty hinged racks, large, for 1000 μl tips
    732-1169
    Log in for availability
    1 * 8 items - 38,00 €
Product Information
  • Catalog #
    Availability
    Price
  • Empty hinged racks, small, for 10 μl and 20 μl tips
    Empty hinged racks, small, for 10 μl and 20 μl tips
    732-1171
    Log in for availability
    1 * 10 items - 43,40 €