Orders LinkContactUsLinkComponent
Pipette tips, ratiolab®
Pipette tips, ratiolab®
Type E, 100 - 1200 µl, -, Transparent, commonly used pipettes, 1000/bag
Catalog # 612-1489
Supplier:   RATIOLAB
Pipette tips, ratiolab®
Catalog # 612-1489
Supplier:  RATIOLAB
Pipette tips, ratiolab®
Pipette tips, ratiolab®
Type E, 100 - 1200 µl, -, Transparent, commonly used pipettes, 1000/bag
Price
13,10 €
Log in for availability
1 * 1.000 items
Product Details & Documents
ratiolab® pipette tips are ideal for use with all commercially available pipettes.
  • Made of high quality PP
  • Tight fit due to the precisely fitting cone
  • Guaranteed dust-free due to special ionization treatment
  • Excellent transparency due to high quality surfaces
  • Minimal residual wetting due to controlled quality of raw materials
  • Excellent volume accuracy due to precise tip orifice
  • MultiTray® refill system is space- and cost-saving and clearly reduces the amount of waste
MultiTray®: Dust-proof refill pack for the Multibox® plus and the Multibox® ultra.
Specifications
  • Packed:
    1000/bag
  • Sterile:
    -
  • Volume:
    100 - 1200 µl
  • Seq:
    26
  • Tip style:
    Type E
  • Tip colour:
    Transparent
  • For use with:
    commonly used pipettes