Orders LinkContactUsLinkComponent
Pipette tips, ratiolab®
Pipette tips, ratiolab®
Type E, 1 - 200 µl, -, Transparent, commonly used pipettes, 1000/bag
  612-1487
 :   RATIOLAB
Pipette tips, ratiolab®
  612-1487
 :  RATIOLAB
Pipette tips, ratiolab®
Pipette tips, ratiolab®
Type E, 1 - 200 µl, -, Transparent, commonly used pipettes, 1000/bag
12,60 €
1 * 1.000 items
ratiolab® pipette tips are ideal for use with all commercially available pipettes.
  • Made of high quality PP
  • Tight fit due to the precisely fitting cone
  • Guaranteed dust-free due to special ionization treatment
  • Excellent transparency due to high quality surfaces
  • Minimal residual wetting due to controlled quality of raw materials
  • Excellent volume accuracy due to precise tip orifice
  • MultiTray® refill system is space- and cost-saving and clearly reduces the amount of waste
MultiTray®: Dust-proof refill pack for the Multibox® plus and the Multibox® ultra.
  • Packed:
    1000/bag
  • Sterile:
    -
  • Volume:
    1 - 200 µl
  • Seq:
    18
  • Tip style:
    Type E
  • Tip colour:
    Transparent
  • For use with:
    commonly used pipettes