Serum pipettes
Serum pipettes, graduated, 2,5 ml, 153 mm, -, 500/inner pack, 1500 pieces/carton
Catalog # 612-1021
Supplier: Greiner Bio-One
Jump to:
Product Details & Documents
These pipettes are suitable for the rapid transfer of liquids.
Specifications
- Packed:500/inner pack, 1500 pieces/carton
- Sterile:-
- Capacity:2,5 ml
- Description:Serum pipettes, graduated
- Length:153 mm
- Seq:18



