Digital Solutions
Applications
Support
About
Export
Orders Link
ContactUsLinkComponent
Bottle Top Dispensers
Accessories
Accessories
Bottle thread adapter, GL 45 to S 40 (Buttress thread), PP
EPPE4960800.147
:
EPPENDORF
Accessories
EPPE4960800.147
:
EPPENDORF
Accessories
Bottle thread adapter, GL 45 to S 40 (Buttress thread), PP
15,30 €
1 * 1 items
-
+
Description:
Bottle thread adapter, GL 45 to S 40 (Buttress thread), PP
Seq:
11