Orders LinkContactUsLinkComponent
Screw caps with liners, WHEATON®
Screw caps with liners, WHEATON®
Screw caps, GPI thread finish 18 - 400, White PP, solid top, Foamed PE
Catalog # 548-2040
Supplier:   DWK Life Sciences
Screw caps with liners, WHEATON®
Catalog # 548-2040
Supplier:  DWK Life Sciences
Screw caps with liners, WHEATON®
Screw caps with liners, WHEATON®
Screw caps, GPI thread finish 18 - 400, White PP, solid top, Foamed PE
Price
82,60 €
Log in for availability
1 * 200 items
Product Details & Documents
These PP screw caps have multi-purpose liners, which are made of three-ply co-extruded foamed low density PE core between two solid layers of low density PE.
  • For use with neutral pH to mild acid and alkali solutions
Specifications
  • Septum:
    Foamed PE
  • Type:
    White PP, solid top
  • Description:
    Screw caps, GPI thread finish 18 - 400
  • Seq:
    01