Test tube baskets
127×102×102 mm
Catalog # 489-2230
Supplier: Bel-Art Products, a Part of SP
Jump to:
Product Details & Documents
PP.
- Very good chemical resistance
- Very good temperature resistance
- Sterilisable and autoclavable
These test tube baskets are designed for rinsing, transporting and storing laboratory glassware.
Colour: White
Specifications
- W×D×H:127×102×102 mm