Order Entry
Export
Orders LinkContactUsLinkComponent
 

Combustion boats

130×19×14 mm
  459-2215
Combustion boats
  459-2215
Combustion boats
Combustion boats

130×19×14 mm

 
39,00 €
   
1 * 1 items
 

Glazed porcelain.

  • With recovery ring
  • Withstand maximum temperature 1000 °C
 
  • Seq:
    03
  • W×D×H:
    130×19×14 mm