Orders LinkContactUsLinkComponent
Lead(II) iodide ≥98.5%
Lead(II) iodide ≥98.5%
Glass bottle for solids, 250 g
  57103.30
Lead(II) iodide ≥98.5%
  57103.30
 :  10101-63-0
Lead(II) iodide ≥98.5%
Lead(II) iodide ≥98.5%
Glass bottle for solids, 250 g
 :  10101-63-0
303,00 €
1 * 250 g
Purity: 98.5% CAS No.: 10101-63-0 Molecular Formula: I2Pb Formula Weight: 461.00 Physical Form: Powder Density: 6.16 Appearance: Yellow Odour: Odourless Melting Point: 402° Boiling Point: 954° Solubility: Partially soluble in water. Freely soluble in sodium thiosulfate solution. Soluble in concentrated solutions of alkali iodides. Insoluble in alcohol and cold HCl Merck Reference: 14,5411 UN No.: 2291 MDL No.: MFCD00011163
 :  Lead diiodide
  •  :10101-63-0
  •  :I₂Pb
  •  :MFCD00011163
  •  :461.01 g/mol
  •  :14,5411
  •  :954 °C
  •  :402 °C
  •  :6.16
  •  :Ambient
  •  :233-256-9
  •  :2291
  •  :6.1,III
  • Pack type:
    Glass bottle for solids
  • Pk:
    250 g