Orders LinkContactUsLinkComponent
Tweezers
Tweezers
250 mm
Catalog # 232-1953
Supplier:   VITLAB
Tweezers
Catalog # 232-1953
Supplier:  VITLAB
Tweezers
Tweezers
250 mm
Price
6,30 €
Log in for availability
1 * 1 items
Product Details & Documents
POM, yellow. Universal tweezers for a variety of applications.
  • Blunt, elastic, very good resilience
  • With grooves on the outside for optimum handling and grip
  • Good chemical resistance
Specifications
  • Length:
    250 mm