Order Entry
Export
Orders LinkContactUsLinkComponent
n-Hexacosane ≥99%

n-Hexacosane ≥99%

Glass bottle for solids, 100 g
Catalog # 42461.22
n-Hexacosane ≥99%
Catalog # 42461.22
CAS Number:  630-01-3
n-Hexacosane ≥99%
n-Hexacosane ≥99%

Glass bottle for solids, 100 g

CAS Number:  630-01-3
Price
218,00 €
Log in for availability
1 * 100 g
Product Details & Documents

Purity: 99% CAS No.: 630-01-3 Molecular Formula: CH3(CH2)24CH3 Formula Weight: 366.72 Physical Form: Solid Appearance: White Odour: Weak Melting Point: 56-58° Beilstein Reference: 1705609 MDL No.: MFCD00009354

Synonyms:  N-hexacosane
  • CAS Number:630-01-3
  • Formula:H₃C(CH₂)₂₄CH₃
  • MDL Number:MFCD00009354
  • Molecular Weight:366.71 g/mol
  • Boiling Pt:412.2 °C (1013 hPa)
  • Melting Pt:56…58 °C
  • Density:0.804 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:211-124-1
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g

Specifications