Orders LinkContactUsLinkComponent
n-Hexacosane ≥99%
Glass bottle for solids, 5 g
  42461.06
n-Hexacosane ≥99%
  42461.06
 :  630-01-3
n-Hexacosane ≥99%
n-Hexacosane ≥99%
Glass bottle for solids, 5 g
 :  630-01-3
40,50 €
1 * 5 g
Purity: 99% CAS No.: 630-01-3 Molecular Formula: CH3(CH2)24CH3 Formula Weight: 366.72 Physical Form: Solid Appearance: White Odour: Weak Melting Point: 56-58° Beilstein Reference: 1705609 MDL No.: MFCD00009354
 :  N-hexacosane
  •  :630-01-3
  •  :H₃C(CH₂)₂₄CH₃
  •  :MFCD00009354
  •  :366.71 g/mol
  •  :412.2 °C (1013 hPa)
  •  :56…58 °C
  •  :0.804 g/cm³ (20 °C)
  •  :Ambient
  •  :211-124-1
  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g