Double spatulas, powder
185 mm, 9 mm
231-1411
: RSG
Stainless steel.
- With round handle, one flat and one scoop shaped, rounded spatula end
- Straight shape
- Very good thermal resistance, autoclavable
- Good chemical resistance
- Length:185 mm
- Blade width:9 mm