Connectors
Fittings, external thread for pipes and valves, 1"
Catalog # 229-0061
Supplier: SEMADENI
Jump to:
Product Details & Documents
Connectors for easy installation on any plastic containers by means of counter screw.
- Male thread for fittings or hose connection
- Good chemical resistance to acids and solvents
Specifications
- Thread:1"
- Description:Fittings, external thread for pipes and valves
- Seq:03