Funnels with short stem
55 mm, 14 mm, 120 mm
221-1156
: SEMADENI
Funnels, made of HDPE, with smooth internal walls. Robust and strong. Multipurpose funnel for a wide variety of uses. Suspension eye for easy storage.
- Easy to clean
- Good chemical resistance
- Good temperature resistance
- Funnel O-Ø:120 mm
- Stem O-Ø:14 mm
- Stem length:55 mm



