Order Entry
Export
Orders LinkContactUsLinkComponent
Funnels with short stem

Funnels with short stem

70 mm, 13 mm, 150 mm
Catalog # 221-1151
Supplier:   SEMADENI
Funnels with short stem
Catalog # 221-1151
Supplier:  SEMADENI
Funnels with short stem
Funnels with short stem

70 mm, 13 mm, 150 mm

Price
7,50 €
Log in for availability
1 * 1 items
Product Details & Documents

Funnels, made of HDPE, with smooth internal walls. Robust and strong. Multipurpose funnel for a wide variety of uses. Suspension eye for easy storage.

  • Easy to clean
  • Good chemical resistance
  • Good temperature resistance
Specifications
  • Funnel O-Ø:
    150 mm
  • Stem O-Ø:
    13 mm
  • Stem length:
    70 mm